siRNA Details
| Gene Symbol: Kif22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009645.1 | NP_001009645.1 | 7 | 1107 | ||
| GenBank mRNA | BC088421.1 | - | 7 | 1107 | 7123493 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif22| Gene Symbol: Kif22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009645.1 | NP_001009645.1 | 7 | 1107 | ||
| GenBank mRNA | BC088421.1 | - | 7 | 1107 | 7123493 | |