siRNA Details
Gene Symbol: Dnah3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759836.1 | XP_008758058.1 | 40, 41 | 6183 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
