siRNA Details
Gene Symbol: Dnah3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759836.1 | XP_008758058.1 | 52 | 10598 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnah3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759836.1 | XP_008758058.1 | 52 | 10598 |