siRNA Details
| Gene Symbol: Npy1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113357.1 | NP_001106828.1 | 2 | 843 | ||
| XM_006253024.2 | XP_006253086.1 | 3 | 1238 | |||
| XM_006253025.2 | XP_006253087.1 | 3 | 1151 | |||
| GenBank mRNA | Z11504.1 | - | 1 | 738 | ||
More Information
Set Membership: |
|

Npy1r
Genomic Map