siRNA Details
| Gene Symbol: Ric8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100520.1 | NP_001093990.1 | 9 | 1675 | ||
| GenBank mRNA | AY177754.1 | - | 9 | 1432 | ||
| BC160852.1 | - | 9 | 1675 | 8364542 | ||
| FQ223245.1 | - | 1370 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ric8a| Gene Symbol: Ric8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100520.1 | NP_001093990.1 | 9 | 1675 | ||
| GenBank mRNA | AY177754.1 | - | 9 | 1432 | ||
| BC160852.1 | - | 9 | 1675 | 8364542 | ||
| FQ223245.1 | - | 1370 | ||||