siRNA Details
| Gene Symbol: Pkp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106315.1 | NP_001099785.1 | 3 | 488 | ||
| XM_006230531.2 | XP_006230593.1 | 2 | 1656 | |||
| XM_008759982.1 | XP_008758204.1 | 2 | 1656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkp3| Gene Symbol: Pkp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106315.1 | NP_001099785.1 | 3 | 488 | ||
| XM_006230531.2 | XP_006230593.1 | 2 | 1656 | |||
| XM_008759982.1 | XP_008758204.1 | 2 | 1656 | |||