siRNA Details
Gene Symbol: LOC100911881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230603.2 | XP_006230665.1 | 736 | |||
XM_006230604.2 | XP_006230666.1 | 762 | ||||
XM_008760034.1 | XP_008758256.1 | 736 | ||||
XM_008760035.1 | XP_008758257.1 | 758 |
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047854.2 | NP_001041319.2 | 7 | 662 | ||
XM_006230548.2 | XP_006230610.1 | 7 | 736 | |||
XM_006230549.2 | XP_006230611.1 | 8 | 762 | |||
GenBank mRNA | AY325242.1 | - | 7 | 634 | ||
BC127546.1 | - | 7 | 662 | 7324297 | ||
FQ213486.1 | - | 7 | 684 | |||
FQ228688.1 | - | 8 | 725 |
