siRNA Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009603.1 | NP_001009603.1 | 2, 3 | 418 | ||
| XM_006230733.2 | XP_006230795.1 | 2, 3 | 427 | |||
| XM_006230734.2 | XP_006230796.1 | 2, 3 | 480 | |||
| XM_008760098.1 | XP_008758320.1 | 2, 3 | 588 | |||
| XM_008760099.1 | XP_008758321.1 | 2, 3 | 480 | |||
| GenBank mRNA | BC088190.1 | - | 2, 3 | 418 | 7371632 | |

Acy3
Genomic Map