siRNA Details
| Gene Symbol: Mpdz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019196.1 | NP_062069.1 | 39 | 5387 | ||
| XM_006238340.2 | XP_006238402.1 | 40 | 5469 | |||
| XM_006238341.2 | XP_006238403.1 | 39 | 5370 | |||
| XM_006238342.2 | XP_006238404.1 | 39 | 5352 | |||
| XM_006238343.2 | XP_006238405.1 | 38 | 5328 | |||
| XM_006238344.2 | XP_006238406.1 | 38 | 5259 | |||
| XM_008763763.1 | XP_008761985.1 | 40 | 6638 | |||
| XM_008763764.1 | XP_008761986.1 | 40 | 5470 | |||
| GenBank mRNA | AJ001320.1 | - | 39 | 5387 | ||

Mpdz
Genomic Map