siRNA Details
| Gene Symbol: Rbm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170484.1 | NP_001163955.1 | 3 | 815 | ||
| XM_008760101.1 | XP_008758323.1 | 3 | 1739 | |||
| GenBank mRNA | FQ212306.1 | - | 3 | 921 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm4| Gene Symbol: Rbm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170484.1 | NP_001163955.1 | 3 | 815 | ||
| XM_008760101.1 | XP_008758323.1 | 3 | 1739 | |||
| GenBank mRNA | FQ212306.1 | - | 3 | 921 | ||