siRNA Details
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106326.1 | NP_001099796.2 | 12 | 1423 | ||
XM_008760105.1 | XP_008758327.1 | 12 | 1411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106326.1 | NP_001099796.2 | 12 | 1423 | ||
XM_008760105.1 | XP_008758327.1 | 12 | 1411 |