siRNA Details
| Gene Symbol: Ctsw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024242.1 | NP_001019413.1 | 7 | 723 | ||
| GenBank mRNA | BC093401.1 | - | 7 | 723 | 7389584 | |
| FQ227585.1 | - | 5 | 522 | |||
| FQ235197.1 | - | 4 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsw| Gene Symbol: Ctsw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024242.1 | NP_001019413.1 | 7 | 723 | ||
| GenBank mRNA | BC093401.1 | - | 7 | 723 | 7389584 | |
| FQ227585.1 | - | 5 | 522 | |||
| FQ235197.1 | - | 4 | 305 | |||