siRNA Details
| Gene Symbol: Ctsw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024242.1 | NP_001019413.1 | 9, 10 | 1013 | ||
| GenBank mRNA | BC093401.1 | - | 9, 10 | 1013 | 7389584 | |
| FQ227585.1 | - | 7, 8 | 812 | |||
| FQ235197.1 | - | 6, 7 | 595 | |||

Ctsw
Genomic Map