siRNA Details
| Gene Symbol: Efemp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005907.2 | NP_001005907.1 | 10 | 1237 | ||
| NM_001277341.1 | NP_001264270.1 | 11 | 1360 | |||
| NM_001277346.1 | NP_001264275.1 | 9 | 1030 | |||
| XM_006230742.2 | XP_006230804.1 | 11 | 1700 | |||
| XM_006230743.1 | XP_006230805.1 | 11 | 1351 | |||
| XM_006230744.2 | XP_006230806.1 | 11 | 1357 | |||
| GenBank mRNA | BC083804.1 | - | 10 | 1175 | 7109182 | |

Efemp2
Genomic Map