siRNA Details
| Gene Symbol: Mus81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025645.1 | NP_001020816.1 | 10 | 1122 | ||
| XM_006230747.2 | XP_006230809.1 | 9 | 1250 | |||
| XM_008760106.1 | XP_008758328.1 | 10 | 1382 | |||
| XM_008760107.1 | XP_008758329.1 | 10 | 1361 | |||
| GenBank mRNA | BC098853.1 | - | 10 | 1122 | 7379711 | |

Mus81
Genomic Map