siRNA Details
| Gene Symbol: Mus81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025645.1 | NP_001020816.1 | 13 | 1498 | ||
| XM_006230747.2 | XP_006230809.1 | 12 | 1626 | |||
| XM_008760106.1 | XP_008758328.1 | 13 | 1758 | |||
| XM_008760107.1 | XP_008758329.1 | 13 | 1737 | |||
| GenBank mRNA | BC098853.1 | - | 13 | 1498 | 7379711 | |

Mus81
Genomic Map