siRNA Details
| Gene Symbol: Cdc42bpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130013.1 | NP_001123485.1 | 5 | 777 | ||
| XM_006230751.2 | XP_006230813.1 | 5 | 716 | |||
| XM_006230752.2 | XP_006230814.1 | 5 | 646 | |||
| XM_008760108.1 | XP_008758330.1 | 5 | 427 | |||

Cdc42bpg
Genomic Map