siRNA Details
| Gene Symbol: Map4k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106329.1 | NP_001099799.1 | 27 | 2248 | ||
| XM_006230753.2 | XP_006230815.1 | 28 | 2219 | |||
| XM_006230754.2 | XP_006230816.1 | 28 | 2192 | |||
| XM_008760109.1 | XP_008758331.1 | 27 | 2072 | |||
| GenBank mRNA | FQ211076.1 | - | 28 | 2189 | ||

Map4k2
Genomic Map