siRNA Details
| Gene Symbol: Hrasls5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039007.1 | NP_001034096.1 | 5 | 1210 | ||
| XM_006230768.2 | XP_006230830.1 | 5 | 942 | |||
| GenBank mRNA | AB255646.1 | - | 5 | 735 | ||
| BC099084.1 | - | 5 | 1210 | 7452977 | ||

Hrasls5
Genomic Map