siRNA Details
| Gene Symbol: Hrasls5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039007.1 | NP_001034096.1 | 3 | 789 | ||
| XM_006230768.2 | XP_006230830.1 | 3 | 521 | |||
| GenBank mRNA | AB255646.1 | - | 3 | 314 | ||
| BC099084.1 | - | 3 | 789 | 7452977 | ||

Hrasls5
Genomic Map