siRNA Details
Gene Symbol: Myrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170487.1 | NP_001163958.1 | 2 | 80 | ||
XM_006231030.2 | XP_006231092.1 | 2 | 241 | |||
XM_006231031.2 | XP_006231093.1 | 2 | 241 | |||
XM_008760220.1 | XP_008758442.1 | 2 | 241 | |||
XM_008760221.1 | XP_008758443.1 | 2 | 241 |
