siRNA Details
| Gene Symbol: Ms4a4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106339.1 | NP_001099809.1 | 2 | 174 | ||
| XM_006231087.2 | XP_006231149.1 | 3 | 343 | |||
| XM_006231088.2 | XP_006231150.1 | 3 | 343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a4c| Gene Symbol: Ms4a4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106339.1 | NP_001099809.1 | 2 | 174 | ||
| XM_006231087.2 | XP_006231149.1 | 3 | 343 | |||
| XM_006231088.2 | XP_006231150.1 | 3 | 343 | |||