siRNA Details
| Gene Symbol: Cct4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182814.2 | NP_877966.1 | 11, 12 | 1385 | ||
| GenBank mRNA | AY223861.1 | - | 11, 12 | 1251 | ||
| BC079283.1 | - | 11, 12 | 1356 | 7134210 | ||
| BQ200856.1 | - | 3, 4 | 575 | |||

Cct4
Genomic Map