siRNA Details
| Gene Symbol: Lpxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009649.1 | NP_001009649.1 | 9 | 1020 | ||
| GenBank mRNA | BC088217.1 | - | 9 | 1020 | 7384057 | |
| FQ230386.1 | - | 9 | 985 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpxn| Gene Symbol: Lpxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009649.1 | NP_001009649.1 | 9 | 1020 | ||
| GenBank mRNA | BC088217.1 | - | 9 | 1020 | 7384057 | |
| FQ230386.1 | - | 9 | 985 | |||