siRNA Details
| Gene Symbol: Acvr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199230.1 | NP_954700.1 | 2 | 293 | ||
| XM_006242315.2 | XP_006242377.1 | 2 | 1137 | |||
| XM_008765678.1 | XP_008763900.1 | 2 | 4298 | |||
| GenBank mRNA | S76466.1 | - | 2 | 293 | ||

Acvr1b
Genomic Map