siRNA Details
| Gene Symbol: Ctbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019201.3 | NP_062074.2 | 6, 7 | 1114 | ||
| XM_006251225.2 | XP_006251287.1 | 5, 6 | 949 | |||
| XM_006251226.2 | XP_006251288.1 | 5, 6 | 949 | |||
| XM_006251227.2 | XP_006251289.1 | 5, 6 | 948 | |||
| XM_006251228.2 | XP_006251290.1 | 5, 6 | 949 | |||
| XM_006251229.2 | XP_006251291.1 | 6, 7 | 1140 | |||
| GenBank mRNA | AF067795.2 | - | 6, 7 | 1119 | ||
| BC078778.1 | - | 6, 7 | 1125 | 7108845 | ||

Ctbp1
Genomic Map