siRNA Details
| Gene Symbol: Bcap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004224.1 | NP_001004224.1 | 4 | 389 | ||
| XM_006229556.2 | XP_006229618.1 | 3 | 346 | |||
| XM_006229557.2 | XP_006229619.1 | 4 | 463 | |||
| GenBank mRNA | BC079182.1 | - | 4 | 389 | 7130880 | |
| FQ219566.1 | - | 4 | 380 | |||
| FQ219790.1 | - | 4 | 388 | |||
| FQ230132.1 | - | 4 | 380 | |||
| FQ230136.1 | - | 4 | 380 | |||

Bcap31
Genomic Map