siRNA Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184045.2 | NP_908934.1 | 3 | 270 | ||
| XM_006229558.2 | XP_006229620.1 | 3 | 387 | |||
| XM_006229559.2 | XP_006229621.1 | 3 | 386 | |||
| XM_006229560.2 | XP_006229622.1 | 3 | 384 | |||
| XM_006229561.2 | XP_006229623.1 | 3 | 384 | |||
| XM_008773617.1 | XP_008771839.1 | 3 | 384 | |||
| XM_008773618.1 | XP_008771840.1 | 3 | 380 | |||
| GenBank mRNA | AY346151.1 | - | 3 | 274 | ||

Srpk3
Genomic Map