siRNA Details
| Gene Symbol: Cxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017183.1 | NP_058879.1 | 2 | 703 | ||
| XM_008767213.1 | XP_008765435.1 | 2 | 1029 | |||
| XM_008767214.1 | XP_008765436.1 | 2 | 1548 | |||
| XM_008767215.1 | XP_008765437.1 | 2 | 869 | |||
| GenBank mRNA | X77797.1 | - | 2 | 703 | ||

Cxcr2
Genomic Map