siRNA Details
| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 1 | 193 | ||
| GenBank mRNA | BC091268.1 | - | 1 | 193 | 7380414 | |
| FQ224897.1 | - | 1 | 228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311863| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 1 | 193 | ||
| GenBank mRNA | BC091268.1 | - | 1 | 193 | 7380414 | |
| FQ224897.1 | - | 1 | 228 | |||