siRNA Details
Gene Symbol: RGD1311863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024974.1 | NP_001020145.1 | 2 | 271 | ||
GenBank mRNA | BC091268.1 | - | 2 | 271 | 7380414 | |
FQ224897.1 | - | 2 | 306 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1311863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024974.1 | NP_001020145.1 | 2 | 271 | ||
GenBank mRNA | BC091268.1 | - | 2 | 271 | 7380414 | |
FQ224897.1 | - | 2 | 306 |