siRNA Details
| Gene Symbol: NEWGENE_1565505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760350.1 | XP_008758572.1 | 3 | 1161 | ||
| XM_008760351.1 | XP_008758573.1 | 3 | 1530 | |||
| XM_008760352.1 | XP_008758574.1 | 2 | 2146 | |||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Human: NM_152629, NM_001042413, DQ438887, DQ438893, DQ438896, and others.
					            	This siRNA also targets the following transcripts in Human: NM_152629, NM_001042413, DQ438887, DQ438893, DQ438896, and others.This siRNA also targets the following transcripts in Mouse: NM_175459, AK054020, DQ871027, AY220846, BC129907, and others.
Panther Classification:
 Back To Top
 Back To Top
 NEWGENE_1565505
NEWGENE_1565505 Genomic Map
Genomic Map Back To Top
 Back To Top