siRNA Details
| Gene Symbol: Bace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019204.2 | NP_062077.1 | 9 | 1895 | ||
| GenBank mRNA | AF190727.1 | - | 9 | 1895 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bace1| Gene Symbol: Bace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019204.2 | NP_062077.1 | 9 | 1895 | ||
| GenBank mRNA | AF190727.1 | - | 9 | 1895 | ||