siRNA Details
| Gene Symbol: Pkd2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106352.1 | NP_001099822.1 | 5 | 1064 | ||
| XM_008760399.1 | XP_008758621.1 | 5 | 899 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkd2l1| Gene Symbol: Pkd2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106352.1 | NP_001099822.1 | 5 | 1064 | ||
| XM_008760399.1 | XP_008758621.1 | 5 | 899 | |||