siRNA Details
Gene Symbol: Smc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106357.1 | NP_001099827.1 | 15 | 2118 | ||
XM_006231183.2 | XP_006231245.1 | 16 | 2256 | |||
XM_006231184.2 | XP_006231246.1 | 16 | 2256 | |||
GenBank mRNA | FQ232653.1 | - | 9 | 1199 |
