siRNA Details
| Gene Symbol: Ccl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019205.1 | NP_062078.1 | 2 | 199 | ||
| GenBank mRNA | U96637.1 | - | 2 | 165 | ||
| Y08358.1 | - | 2 | 199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl11| Gene Symbol: Ccl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019205.1 | NP_062078.1 | 2 | 199 | ||
| GenBank mRNA | U96637.1 | - | 2 | 165 | ||
| Y08358.1 | - | 2 | 199 | |||