siRNA Details
Gene Symbol: Kazald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033064.1 | NP_001028236.1 | 2 | 178 | ||
GenBank mRNA | BC103652.1 | - | 2 | 178 | 7317353 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kazald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033064.1 | NP_001028236.1 | 2 | 178 | ||
GenBank mRNA | BC103652.1 | - | 2 | 178 | 7317353 |