siRNA Details
| Gene Symbol: Obfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011943.1 | NP_001011943.1 | 7 | 846 | ||
| XM_006231472.1 | XP_006231534.1 | 7 | 957 | |||
| XM_006231474.2 | XP_006231536.1 | 7 | 955 | |||
| XM_006231475.2 | XP_006231537.1 | 5 | 3289 | |||
| GenBank mRNA | BC079096.1 | - | 7 | 846 | 7124109 | |

Obfc1
Genomic Map