siRNA Details
| Gene Symbol: Dntt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012461.1 | NP_001012479.1 | 9, 10 | 1373 | ||
| GenBank mRNA | BC089904.1 | - | 9, 10 | 1373 | 7384335 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dntt| Gene Symbol: Dntt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012461.1 | NP_001012479.1 | 9, 10 | 1373 | ||
| GenBank mRNA | BC089904.1 | - | 9, 10 | 1373 | 7384335 | |