siRNA Details
| Gene Symbol: Kcnv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106370.1 | NP_001099840.1 | 2 | 1752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnv2| Gene Symbol: Kcnv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106370.1 | NP_001099840.1 | 2 | 1752 | ||