siRNA Details
Gene Symbol: Ffar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047088.1 | NP_001040553.1 | 3 | 1107 | ||
GenBank mRNA | AB207868.1 | - | 3 | 1107 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
