siRNA Details
| Gene Symbol: Ifit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024753.1 | NP_001019924.1 | 2 | 993 | ||
| GenBank mRNA | BC097384.1 | - | 2 | 993 | 7446350 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifit2| Gene Symbol: Ifit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024753.1 | NP_001019924.1 | 2 | 993 | ||
| GenBank mRNA | BC097384.1 | - | 2 | 993 | 7446350 | |