siRNA Details
| Gene Symbol: Lipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106374.1 | NP_001099844.1 | 7 | 805 | ||
| XM_008760325.1 | XP_008758547.1 | 8 | 813 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipk| Gene Symbol: Lipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106374.1 | NP_001099844.1 | 7 | 805 | ||
| XM_008760325.1 | XP_008758547.1 | 8 | 813 | |||