siRNA Details
| Gene Symbol: Adra1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017191.2 | NP_058887.2 | 1 | 762 | ||
| XM_006252107.2 | XP_006252169.1 | 2 | 1747 | |||
| XM_006252108.2 | XP_006252170.1 | 2 | 1745 | |||
| XM_006252109.2 | XP_006252171.1 | 2 | 1746 | |||
| XM_006252110.2 | XP_006252172.1 | 2 | 1741 | |||
| XM_008770744.1 | XP_008768966.1 | 2 | 1747 | |||
| GenBank mRNA | S67375.1 | - | 1 | 159 | ||
| U07126.1 | - | 1 | 762 | |||
| U13368.1 | - | 1 | 782 | |||
More Information
Set Membership: |
|

Adra1a
Genomic Map