siRNA Details
| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031575.1 | NP_113763.1 | 10 | 1047 | ||
| GenBank mRNA | D49836.1 | - | 10 | 1047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akt3| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031575.1 | NP_113763.1 | 10 | 1047 | ||
| GenBank mRNA | D49836.1 | - | 10 | 1047 | ||