siRNA Details
| Gene Symbol: Olr1688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000275.1 | NP_001000275.1 | 767 | |||
| Gene Symbol: LOC100910382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751905.2 | XP_003751953.1 | 1 | 780 | ||

Olr1688
Genomic Map