siRNA Details
| Gene Symbol: Aadat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017193.1 | NP_058889.1 | 1, 2 | 174 | ||
| GenBank mRNA | BC078864.1 | - | 1, 2 | 170 | 7109210 | |
| Z50144.1 | - | 1, 2 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aadat| Gene Symbol: Aadat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017193.1 | NP_058889.1 | 1, 2 | 174 | ||
| GenBank mRNA | BC078864.1 | - | 1, 2 | 170 | 7109210 | |
| Z50144.1 | - | 1, 2 | 174 | |||