siRNA Details
| Gene Symbol: Aadat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017193.1 | NP_058889.1 | 8, 9 | 1019 | ||
| XM_006253067.2 | XP_006253129.1 | 8, 9 | 1107 | |||
| GenBank mRNA | BC078864.1 | - | 8, 9 | 1015 | 7109210 | |
| Z50144.1 | - | 8, 9 | 1019 | |||

Aadat
Genomic Map