siRNA Details
| Gene Symbol: Dhx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212496.2 | NP_997661.1 | 18, 19 | 2971 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhx16| Gene Symbol: Dhx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212496.2 | NP_997661.1 | 18, 19 | 2971 | ||