siRNA Details
| Gene Symbol: Gtf2h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212501.2 | NP_997666.1 | 13 | 1338 | ||
| XM_006255920.2 | XP_006255982.1 | 13 | 1387 | |||
| GenBank mRNA | BC083703.1 | - | 11 | 1564 | ||
| BC127469.1 | - | 13 | 1310 | 7324889 | ||

Gtf2h4
Genomic Map